Neutec Group SAL-T Humidity Standards
Humidity standards based on saturated salt solutions in plastic tablets with humidity-permeable membrane for multi-use. The salts generate a defined air humidity in the temperature range of 15° to 30°C which flows out of the tablet through the permeable membrane inside the measuring chamber and generate there a defined and stable humidity environment.
Supplier: Neutec Group NV1110936
Description
- For verification and calibration of Novasina water activity meters
- Precise and high repeatability
- Includes an internationally recognized certificate from an accredited European laboratory
- Multi applicable
- High accuracy and repeatability
- Simple handling
- Available for complete measuring range
Specifications
SAL-T 75% |