Neutec Group SAL-T Humidity Standards
Humidity standards based on saturated salt solutions in plastic tablets with humidity-permeable membrane for multi-use. The salts generate a defined air humidity in the temperature range of 15° to 30°C which flows out of the tablet through the permeable membrane inside the measuring chamber and generate there a defined and stable humidity environment.
$655.15 - $655.15
Products 1
Catalog Number Mfr. No. Description Price Quantity  
Catalog Number Mfr. No. Description Price Quantity  
14-910-402
View Documents
Neutec Group
NV1110936
SAL-T 75%
Each for $655.15
Description
Description
  • For verification and calibration of Novasina water activity meters
  • Precise and high repeatability
  • Includes an internationally recognized certificate from an accredited European laboratory
  • Multi applicable
  • High accuracy and repeatability
  • Simple handling
  • Available for complete measuring range
Videos
SDS
missing translation for 'documents'
missing translation for 'documents'