missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Salbutamol hemisulfate, 98+%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals A1854414

 View more versions of this product

Catalog No. AAA1854414


Explore available promotions

Description

Description

Salbutamol sulfate is used as standard in analysis of fortified meat for LABA content using ELISA kits. It is also used to study the response of prostaglandin 2α to various β-adrenergic agonists in cultured bovine endometrial cells.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Salbutamol sulfate is used as standard in analysis of fortified meat for LABA content using ELISA kits. It is also used to study the response of prostaglandin 2α to various β-adrenergic agonists in cultured bovine endometrial cells.

Solubility
Soluble in water and 1M sodium hydroxide (50 mg/ml).

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
Specifications

Specifications

Salbutamol sulfate
C13H23NO7S
25 g
salbutamol sulfate, albuterol sulfate, ventolin, ventorlin, 4-2-tert-butylamino-1-hydroxyethyl-2-hydroxymethyl phenol sulfate, asthavent, albuterol sulfate 1:x, salbutamol; sulfuric acid, asmaven sulfate, 2-tert-butylamino-1-4-hydroxy-3-hydroxymethylphenyl ethanol hemisulfate
OVICLFZZVQVVFT-UHFFFAOYSA-N
4-[2-(tert-butylamino)-1-hydroxyethyl]-2-(hydroxymethyl)phenol;sulfuric acid
9884233
99%
51022-70-9
MFCD00055200
Light sensitive
Soluble in water and 1M sodium hydroxide (50mg/ml).
CC(C)(C)NCC(C1=CC(=C(C=C1)O)CO)O.OS(=O)(=O)O
337.387
576.7

Chemical Identifiers

51022-70-9
337.387
OVICLFZZVQVVFT-UHFFFAOYSA-N
9884233
CC(C)(C)NCC(C1=CC(=C(C=C1)O)CO)O.OS(=O)(=O)O
C13H23NO7S
MFCD00055200
salbutamol sulfate, albuterol sulfate, ventolin, ventorlin, 4-2-tert-butylamino-1-hydroxyethyl-2-hydroxymethyl phenol sulfate, asthavent, albuterol sulfate 1:x, salbutamol; sulfuric acid, asmaven sulfate, 2-tert-butylamino-1-4-hydroxy-3-hydroxymethylphenyl ethanol hemisulfate
4-[2-(tert-butylamino)-1-hydroxyethyl]-2-(hydroxymethyl)phenol;sulfuric acid
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H302
Harmful if swallowed.

P201-P202-P261-P272-P280g-P281-P302+P352-P308+P313-P333+P313-P363-P501c

H317-H351-H361fd

EINECSNumber : 256-916-8

RTECSNumber : CZ6430100

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only