Learn More
Salbutamol hemisulfate, 98+%, Thermo Scientific Chemicals
$95.36 - $1182.03
Chemical Identifiers
CAS | 51022-70-9 |
---|---|
Molecular Formula | C13H23NO7S |
Molecular Weight (g/mol) | 337.387 |
MDL Number | MFCD00055200 |
InChI Key | OVICLFZZVQVVFT-UHFFFAOYSA-N |
Synonym | salbutamol sulfate, albuterol sulfate, ventolin, ventorlin, 4-2-tert-butylamino-1-hydroxyethyl-2-hydroxymethyl phenol sulfate, asthavent, albuterol sulfate 1:x, salbutamol; sulfuric acid, asmaven sulfate, 2-tert-butylamino-1-4-hydroxy-3-hydroxymethylphenyl ethanol hemisulfate |
PubChem CID | 9884233 |
IUPAC Name | 4-[2-(tert-butylamino)-1-hydroxyethyl]-2-(hydroxymethyl)phenol;sulfuric acid |
SMILES | CC(C)(C)NCC(C1=CC(=C(C=C1)O)CO)O.OS(=O)(=O)O |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAA1854403
|
Thermo Scientific Chemicals
A1854403 |
1 g |
Each for $95.36
|
|
|||||
AAA1854406
|
Thermo Scientific Chemicals
A1854406 |
5 g |
Each for $346.60
|
|
|||||
AAA1854414
|
Thermo Scientific Chemicals
A1854414 |
25 g |
Each for $1,182.03
|
|
|||||
Description
Salbutamol sulfate is used as standard in analysis of fortified meat for LABA content using ELISA kits. It is also used to study the response of prostaglandin 2α to various β-adrenergic agonists in cultured bovine endometrial cells.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsSalbutamol sulfate is used as standard in analysis of fortified meat for LABA content using ELISA kits. It is also used to study the response of prostaglandin 2α to various β-adrenergic agonists in cultured bovine endometrial cells.
Solubility
Soluble in water and 1M sodium hydroxide (50 mg/ml).
Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
Chemical Identifiers
51022-70-9 | |
337.387 | |
OVICLFZZVQVVFT-UHFFFAOYSA-N | |
9884233 | |
CC(C)(C)NCC(C1=CC(=C(C=C1)O)CO)O.OS(=O)(=O)O |
C13H23NO7S | |
MFCD00055200 | |
salbutamol sulfate, albuterol sulfate, ventolin, ventorlin, 4-2-tert-butylamino-1-hydroxyethyl-2-hydroxymethyl phenol sulfate, asthavent, albuterol sulfate 1:x, salbutamol; sulfuric acid, asmaven sulfate, 2-tert-butylamino-1-4-hydroxy-3-hydroxymethylphenyl ethanol hemisulfate | |
4-[2-(tert-butylamino)-1-hydroxyethyl]-2-(hydroxymethyl)phenol;sulfuric acid |
Specifications
51022-70-9 | |
MFCD00055200 | |
Light sensitive | |
Soluble in water and 1M sodium hydroxide (50mg/ml). | |
CC(C)(C)NCC(C1=CC(=C(C=C1)O)CO)O.OS(=O)(=O)O | |
337.387 | |
576.7 | |
Salbutamol sulfate |
C13H23NO7S | |
1 g | |
salbutamol sulfate, albuterol sulfate, ventolin, ventorlin, 4-2-tert-butylamino-1-hydroxyethyl-2-hydroxymethyl phenol sulfate, asthavent, albuterol sulfate 1:x, salbutamol; sulfuric acid, asmaven sulfate, 2-tert-butylamino-1-4-hydroxy-3-hydroxymethylphenyl ethanol hemisulfate | |
OVICLFZZVQVVFT-UHFFFAOYSA-N | |
4-[2-(tert-butylamino)-1-hydroxyethyl]-2-(hydroxymethyl)phenol;sulfuric acid | |
9884233 | |
99% |
Safety and Handling
GHS H Statement
H302
Harmful if swallowed.
P201-P202-P261-P272-P280g-P281-P302+P352-P308+P313-P333+P313-P363-P501c
H317-H351-H361fd
EINECSNumber : 256-916-8
RTECSNumber : CZ6430100
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only