missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Salicylaldehyde, 99%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 132602500

 View more versions of this product

Catalog No. AC132602500


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Salicylaldehyde
-7.0°C
1.1600g/mL
76°C
1% max. (Phenol) (GC)
C7H6O2
2-(HO)C6H4CHO
250 g
1.16
salicylaldehyde, o-hydroxybenzaldehyde, o-formylphenol, salicylal, 2-formylphenol, benzaldehyde, 2-hydroxy, salicylic aldehyde, salicyladehyde, 2-hydroxy-benzaldehyde, benzaldehyde, o-hydroxy
SMQUZDBALVYZAC-UHFFFAOYSA-N
2-hydroxybenzaldehyde
6998
122.12
Liquid
90-02-8,108-95-2
Yellow
197.0°C
Authentic
Glass bottle
1.5720 to 1.5740
MFCD00003317
08,31
15,8462
Solubility in water: slightly soluble. Other solubilities: soluble in alcohol, ether and benzene
OC1=CC=CC=C1C=O
122.12
CHEBI:16008
99%

Chemical Identifiers

90-02-8
122.12
SMQUZDBALVYZAC-UHFFFAOYSA-N
6998
2-hydroxybenzaldehyde
C7H6O2
MFCD00003317
salicylaldehyde, o-hydroxybenzaldehyde, o-formylphenol, salicylal, 2-formylphenol, benzaldehyde, 2-hydroxy, salicylic aldehyde, salicyladehyde, 2-hydroxy-benzaldehyde, benzaldehyde, o-hydroxy
CHEBI:16008
OC1=CC=CC=C1C=O
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Harmful if swallowed.
Toxic in contact with skin.
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement
Wash face,hands and any exposed skin thoroughly after handling.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
IF ON SKIN: Wash with p

GHS Signal Word: Danger

EINECSNumber : 201-961-

Recommended Storage : product darkens during storage

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only