missing translation for 'onlineSavingsMsg'
Learn More

Salicylaldehyde, 99%

CAS: 90-02-8 | C7H6O2 | 122.12 g/mol

$67.20 - $107.86

Chemical Identifiers

CAS 90-02-8
Molecular Formula C7H6O2
Molecular Weight (g/mol) 122.12
MDL Number MFCD00003317
InChI Key SMQUZDBALVYZAC-UHFFFAOYSA-N
Synonym salicylaldehyde, o-hydroxybenzaldehyde, o-formylphenol, salicylal, 2-formylphenol, benzaldehyde, 2-hydroxy, salicylic aldehyde, salicyladehyde, 2-hydroxy-benzaldehyde, benzaldehyde, o-hydroxy
PubChem CID 6998
ChEBI CHEBI:16008
IUPAC Name 2-hydroxybenzaldehyde
SMILES OC1=CC=CC=C1C=O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC132601000
SDS View Documents
Thermo Scientific Chemicals
132601000
100 g Glass bottle
Each for $67.20
Only null left
 
AC132602500
SDS View Documents
Thermo Scientific Chemicals
132602500
250 g Glass bottle
Each for $107.86
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

90-02-8
122.12
SMQUZDBALVYZAC-UHFFFAOYSA-N
6998
2-hydroxybenzaldehyde
C7H6O2
MFCD00003317
salicylaldehyde, o-hydroxybenzaldehyde, o-formylphenol, salicylal, 2-formylphenol, benzaldehyde, 2-hydroxy, salicylic aldehyde, salicyladehyde, 2-hydroxy-benzaldehyde, benzaldehyde, o-hydroxy
CHEBI:16008
OC1=CC=CC=C1C=O

Specifications

90-02-8,108-95-2
Yellow
197.0°C
Authentic
Glass bottle
1.5720 to 1.5740
MFCD00003317
08,31
15,8462
Solubility in water: slightly soluble. Other solubilities: soluble in alcohol, ether and benzene
OC1=CC=CC=C1C=O
122.12
CHEBI:16008
99%
Salicylaldehyde
-7.0°C
1.1600g/mL
76°C
1% max. (Phenol) (GC)
C7H6O2
2-(HO)C6H4CHO
100 g
1.16
salicylaldehyde, o-hydroxybenzaldehyde, o-formylphenol, salicylal, 2-formylphenol, benzaldehyde, 2-hydroxy, salicylic aldehyde, salicyladehyde, 2-hydroxy-benzaldehyde, benzaldehyde, o-hydroxy
SMQUZDBALVYZAC-UHFFFAOYSA-N
2-hydroxybenzaldehyde
6998
122.12
Liquid
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Harmful if swallowed.
Toxic in contact with skin.
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement
Wash face,hands and any exposed skin thoroughly after handling.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
IF ON SKIN: Wash with p

GHS Signal Word: Danger

EINECSNumber : 201-961-

Recommended Storage : product darkens during storage

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only