missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Salicylic acid, sodium salt, 99+%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 220982500

 View more versions of this product

Catalog No. AC220982500


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Salicylic acid, sodium salt
2mL 0.01N NaOH max.
Brown to Brown-Red
Authentic
Plastic bottle
2-(HO)C6H4CO2Na
250 g
15, 8469
Solubility in water: 1000g/L (20°C). Other solubilities: 1g/9.2mL alcohol, 1g/4mL glycerol, practically insoluble in ether
C1=CC=C(C(=C1)C(=O)[O-])O.[Na+]
160.104
CHEBI:9180
≥99%
54-21-7
200.0°C
0.5% max. (1g, 105°C)
99+%
C7H5NaO3
MFCD00002440
10, 43
sodium salicylate, sodium 2-hydroxybenzoate, salsonin, clin, enterosalicyl, enterosalil, entrosalyl, glutosalyl, kerasalicyl, magsalyl
ABBQHOQBGMUPJH-UHFFFAOYSA-M
sodium;2-hydroxybenzoate
16760658
160.1
Crystalline Powder or Crystals

Chemical Identifiers

54-21-7
160.104
ABBQHOQBGMUPJH-UHFFFAOYSA-M
16760658
sodium;2-hydroxybenzoate
C7H5NaO3
MFCD00002440
sodium salicylate, sodium 2-hydroxybenzoate, salsonin, clin, enterosalicyl, enterosalil, entrosalyl, glutosalyl, kerasalicyl, magsalyl
CHEBI:9180
C1=CC=C(C(=C1)C(=O)[O-])O.[Na+]
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Harmful if swallowed.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only