Learn More
Salicylic acid, sodium salt, 99+%, Thermo Scientific Chemicals
$91.30 - $1,610.96
Chemical Identifiers
CAS | 54-21-7 |
---|---|
Molecular Formula | C7H5NaO3 |
Molecular Weight (g/mol) | 160.104 |
MDL Number | MFCD00002440 |
InChI Key | ABBQHOQBGMUPJH-UHFFFAOYSA-M |
Synonym | sodium salicylate, sodium 2-hydroxybenzoate, salsonin, clin, enterosalicyl, enterosalil, entrosalyl, glutosalyl, kerasalicyl, magsalyl |
PubChem CID | 16760658 |
ChEBI | CHEBI:9180 |
IUPAC Name | sodium;2-hydroxybenzoate |
SMILES | C1=CC=C(C(=C1)C(=O)[O-])O.[Na+] |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AC220980500
|
Thermo Scientific Chemicals
220980500 |
50 g | Plastic bottle |
N/A
|
|
||||
AC220982500
|
Thermo Scientific Chemicals
220982500 |
250 g | Plastic bottle |
N/A
|
|
||||
AC220980010
|
Thermo Scientific Chemicals
220980010 |
1 kg | Plastic bottle |
N/A
|
|
||||
AC220980025
|
Thermo Scientific Chemicals
220980025 |
2.5 kg | Plastic bottle |
N/A
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
54-21-7 | |
160.104 | |
ABBQHOQBGMUPJH-UHFFFAOYSA-M | |
16760658 | |
sodium;2-hydroxybenzoate |
C7H5NaO3 | |
MFCD00002440 | |
sodium salicylate, sodium 2-hydroxybenzoate, salsonin, clin, enterosalicyl, enterosalil, entrosalyl, glutosalyl, kerasalicyl, magsalyl | |
CHEBI:9180 | |
C1=CC=C(C(=C1)C(=O)[O-])O.[Na+] |
Specifications
54-21-7 | |
200.0°C | |
0.5% max. (1g, 105°C) | |
99+% | |
C7H5NaO3 | |
MFCD00002440 | |
10, 43 | |
sodium salicylate, sodium 2-hydroxybenzoate, salsonin, clin, enterosalicyl, enterosalil, entrosalyl, glutosalyl, kerasalicyl, magsalyl | |
ABBQHOQBGMUPJH-UHFFFAOYSA-M | |
sodium;2-hydroxybenzoate | |
16760658 | |
160.1 | |
Crystalline Powder or Crystals |
2mL 0.01N NaOH max. | |
Brown to Brown-Red | |
Authentic | |
Plastic bottle | |
2-(HO)C6H4CO2Na | |
50 g | |
15, 8469 | |
Solubility in water: 1000g/L (20°C). Other solubilities: 1g/9.2mL alcohol, 1g/4mL glycerol, practically insoluble in ether | |
C1=CC=C(C(=C1)C(=O)[O-])O.[Na+] | |
160.104 | |
CHEBI:9180 | |
≥99% | |
Salicylic acid, sodium salt |
Safety and Handling
GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Harmful if swallowed.
Causes serious eye irritation.
GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
RUO – Research Use Only