Thermo Scientific™ Samco™ Special Purpose Transfer Pipettes
Eliminate plugging, packaging and autoclaving with these special purpose transfer pipettes, ideal for delivering drops onto slides or tiles.
$70.59 - $120.96
Catalog Number | Mfr. No. | Volume (Metric) | Sterility | Bulb Draw (Metric) | Description | Price | Quantity | ||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Volume (Metric) | Sterility | Bulb Draw (Metric) | Description | Price | Quantity | ||
13-711-44
|
Thermo Scientific™
292 |
1.0 mL | Non-sterile | 0.2 mL | Padl-Pet 25μL Drop Size |
Pack of 500 Each for $120.96
Case of 8 Pack for $967.68
|
|
||
13-711-45
|
Thermo Scientific™
293 |
1.0 mL | Non-sterile | 0.2 mL | Padl-Pet 30μL Drop Size |
Pack of 500 Each for $95.50
Case of 8 Pack for $764.00
|
|
||
13-711-46
|
Thermo Scientific™
295 |
0.8 mL | Non-sterile | 0.2 mL | Padl-Pet 50μL Drop Size |
Pack of 500 Each for $70.59
Case of 8 Pack for $564.72
|
|
||
Description
Eliminate plugging, packaging and autoclaving with Thermo Scientific™ Samco™ Special Purpose Transfer Pipettes, ideal for delivering drops onto slides or tiles. Single-unit design features a built-in pipettes bulb that eliminates the need for plugging, packaging, autoclaving; prevents cross-contamination.
- Shatterproof, nontoxic polyethylene
- Low-affinity surfaces –prevents proteins binding in research applications
- Can be frozen in liquid nitrogen, or heat sealed for storage
- Nonsterile
Specifications
LDPE |