missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Sand, pure, 40-100 mesh, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 370940010
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Filter aidSpecifications
2230.0°C | |
1610.0°C | |
Beige to Gray | |
Crystals | |
O2Si | |
MFCD00011232,MFCD00217788,MFCD00163736,MFCD07370733 | |
silica, silicon dioxide, quartz, cristobalite, diatomaceous earth, tridymite, silicic anhydride, aerosil, infusorial earth, silica, amorphous | |
VYPSYNLAJGMNEJ-UHFFFAOYSA-N | |
silanedione | |
24261 | |
60.08 | |
Plastic bottle |
Sand | |
40 to 100 Mesh | |
14808-60-7 | |
1 kg | |
SiO2 | |
13,8567 | |
Solubility in water: insoluble. | |
O=[Si]=O | |
60.08 | |
CHEBI:30563 | |
Pure |
Chemical Identifiers
14808-60-7 | |
60.08 | |
VYPSYNLAJGMNEJ-UHFFFAOYSA-N | |
24261 | |
O=[Si]=O |
O2Si | |
MFCD00011232,MFCD00217788,MFCD00163736,MFCD07370733 | |
silica, silicon dioxide, quartz, cristobalite, diatomaceous earth, tridymite, silicic anhydride, aerosil, infusorial earth, silica, amorphous | |
CHEBI:30563 |
Safety and Handling
EINECSNumber : 238-878-4
RUO – Research Use Only