missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Sand, pure, 40-100 mesh, Thermo Scientific Chemicals
$58.12 - $857.33
Chemical Identifiers
CAS | 14808-60-7 |
---|---|
Molecular Formula | O2Si |
Molecular Weight (g/mol) | 60.08 |
MDL Number | MFCD00011232,MFCD00217788,MFCD00163736,MFCD07370733 |
InChI Key | VYPSYNLAJGMNEJ-UHFFFAOYSA-N |
Synonym | silica, silicon dioxide, quartz, cristobalite, diatomaceous earth, tridymite, silicic anhydride, aerosil, infusorial earth, silica, amorphous |
PubChem CID | 24261 |
ChEBI | CHEBI:30563 |
SMILES | O=[Si]=O |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | |||||
AC370942500
|
Thermo Scientific Chemicals
370942500 |
250 g | Plastic bottle |
Each for $58.12
|
|
|||||
AC370940010
|
Thermo Scientific Chemicals
370940010 |
1 kg | Plastic bottle |
Each for $92.69
|
|
|||||
AC370940050
|
Thermo Scientific Chemicals
370940050 |
5 kg | Plastic bottle |
Each for $253.64
|
|
|||||
AC370940250
|
Thermo Scientific Chemicals
370940250 |
25 kg | Fiber Drum |
Each for $857.33
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Filter aidChemical Identifiers
14808-60-7 | |
60.08 | |
VYPSYNLAJGMNEJ-UHFFFAOYSA-N | |
24261 | |
O=[Si]=O |
O2Si | |
MFCD00011232,MFCD00217788,MFCD00163736,MFCD07370733 | |
silica, silicon dioxide, quartz, cristobalite, diatomaceous earth, tridymite, silicic anhydride, aerosil, infusorial earth, silica, amorphous | |
CHEBI:30563 |
Specifications
2230.0°C | |
Beige to Gray | |
Crystals | |
O2Si | |
MFCD00011232,MFCD00217788,MFCD00163736,MFCD07370733 | |
silica, silicon dioxide, quartz, cristobalite, diatomaceous earth, tridymite, silicic anhydride, aerosil, infusorial earth, silica, amorphous | |
VYPSYNLAJGMNEJ-UHFFFAOYSA-N | |
dioxosilane | |
24261 | |
60.08 | |
Plastic bottle |
1610.0°C | |
14808-60-7 | |
250 g | |
SiO2 | |
13,8567 | |
Solubility in water: insoluble. | |
O=[Si]=O | |
60.08 | |
CHEBI:30563 | |
Pure | |
Sand, 40 to 100 Mesh |
Safety and Handling
EINECSNumber : 238-878-4
RUO – Research Use Only