missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Scandium(III) oxide, 99.999%, (trace metal basis), Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 193960050
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
for ultra trace analysisSpecifications
Scandium(III) oxide | |
White | |
Powder | |
99.999% | |
SC2O3 | |
10,8239 | |
HJGMWXTVGKLUAQ-UHFFFAOYSA-N | |
discandium(3+) trioxidandiide | |
137.91 | |
Trace Metal Basis |
>1000.0°C | |
12060-08-1 | |
5 g | |
O3Sc2 | |
MFCD00011223 | |
Solubility in water: insoluble | |
[O--].[O--].[O--].[Sc+3].[Sc+3] | |
137.91 | |
99.999% | |
Glass bottle |
Chemical Identifiers
12060-08-1 | |
137.91 | |
HJGMWXTVGKLUAQ-UHFFFAOYSA-N | |
[O--].[O--].[O--].[Sc+3].[Sc+3] |
O3Sc2 | |
MFCD00011223 | |
discandium(3+) trioxidandiide |
Safety and Handling
EINECSNumber : 235-042-
RUO – Research Use Only