missing translation for 'onlineSavingsMsg'
Learn More

Scandium(III) oxide, 99.999%, (trace metal basis), Thermo Scientific Chemicals

$465.73 - $465.73

Chemical Identifiers

CAS 12060-08-1
Molecular Formula O3Sc2
Molecular Weight (g/mol) 137.91
MDL Number MFCD00011223
InChI Key HJGMWXTVGKLUAQ-UHFFFAOYSA-N
IUPAC Name discandium(3+) trioxidandiide
SMILES [O--].[O--].[O--].[Sc+3].[Sc+3]
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC193960050
SDS View Documents
Thermo Scientific Chemicals
193960050
5 g Glass bottle
Each for $465.73
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

for ultra trace analysis
Specifications

Chemical Identifiers

12060-08-1
137.91
HJGMWXTVGKLUAQ-UHFFFAOYSA-N
[O--].[O--].[O--].[Sc+3].[Sc+3]
O3Sc2
MFCD00011223
discandium(3+) trioxidandiide

Specifications

>1000.0°C
12060-08-1
5 g
O3Sc2
MFCD00011223
Solubility in water: insoluble
[O--].[O--].[O--].[Sc+3].[Sc+3]
137.91
99.999%
Glass bottle
White
Powder
99.999%
SC2O3
10,8239
HJGMWXTVGKLUAQ-UHFFFAOYSA-N
discandium(3+) trioxidandiide
137.91
Trace Metal Basis
Scandium(III) oxide
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 235-042-

SDS
Documents

Documents

RUO – Research Use Only