Ampac™ SealPAK Tubullar Rollstock Pouches
Create custom-size pouches
$1468.71 - $3609.63
Catalog Number | Mfr. No. | Thickness (English) | Width (Metric) | Material | Price | Quantity | |||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Thickness (English) | Width (Metric) | Material | Price | Quantity | |||
01-812-26C
|
Ampac™
TRS242506 |
2.50 mil | 61 cm | Polyester, Polyethylene |
Each for $3,609.63
|
|
|||
01-812-26D
|
Ampac™
TRS951253 |
4.50 mil | 24 cm | Polyester, Polyethylene |
Each for $2,034.67
|
|
|||
01-812-26A
|
Ampac™
TRS161254 |
4.50 mil | 41 cm | Polyester, Polyethylene |
Each for $3,065.07
|
|
|||
01-812-26E
|
Ampac™
TRS952502 |
2.50 mil | 24 cm | Polyester, Polyethylene |
Each for $2,141.77
|
|
|||
01-812-782
|
Ampac™
TRSF951253 |
4.50 mil | 24 cm | Foil |
Each for $1,468.71
|
|
|||
Description
Create custom-size pouches
- Ideal for histology and pathology specimens and for safe biological waste disposal in medical, institutional and industrial facilities
- Polyester with polyethylene lining; does not react with formaldehyde, weak solutions of acids, alkalis or organic solvents
- Withstand infrared and gamma rays
- Can be frozen or boiled
- Can be cut to desired length and sealed at both ends
Compliance
Use no adhesives or bonding agents.
Specifications
Heat-Sealable Roll Stock Pouches |