3M™ Secure Click™ Half Facepiece Reusable Respirator with Speaking Diaphragm
This Half Facepiece Reusable Respirator comes with a speaking diaphragm which is designed to help provide easier communication while you work.
Supplier:  3M™ HF802SD
 View more versions of this product
Catalog No. 17-100-814
Explore available promotions
Description
Description
  • Designed with smart and intuitive features
  • Simple, comfortable and reliable
  • Quick, easy, one-touch negative pressure push button seal check provides improved confidence in respirator fit
Specifications
Specifications
Black
Reusable
Respirator
Yes
10 Respirators/Pk.
Half Facepiece
Silicone, Nylon
Medium
Yes