missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
3M™ Secure Click™ Half Facepiece Reusable Respirator with Speaking Diaphragm
This Half Facepiece Reusable Respirator comes with a speaking diaphragm which is designed to help provide easier communication while you work.
$590.59 - $590.59
Specifications
Color | Black |
---|---|
Design Type | Half Facepiece |
Disposable | Reusable |
Material | Silicone, Nylon |
Product Type | Respirator |
Catalog Number | Mfr. No. | Size | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Size | Price | Quantity | |||||
17-100-814
|
3M™
HF802SD |
Medium |
Case of 10 Each for $590.59
|
|
|||||
Description
- Designed with smart and intuitive features
- Simple, comfortable and reliable
- Quick, easy, one-touch negative pressure push button seal check provides improved confidence in respirator fit
Specifications
Black | |
Reusable | |
Respirator | |
Yes |
Half Facepiece | |
Silicone, Nylon | |
Yes | |
10 Respirators/Pk. |