3M™ SecureFit™ 3700 Series Protective Eyewear
Get over-the-glass protection with all day wearability
$163.62 - $163.62
Specifications
Product Type Protective eyewear
For Use With (Application) Automotive, Machining, Transportation, Sawing, Riveting
No. per Case 10/Cs.
Size Universal
Temple Style 3M™ Pressure Diffusion Temple Technology
View More Specs
Products 2
Catalog Number Mfr. No. Lens Tint Lens Coating Temple Color Description Price Quantity  
Catalog Number Mfr. No. Lens Tint Lens Coating Temple Color Description Price Quantity  
19-071-160
View Documents
3M™
7100203179
Clear Anti-fog Blue Regular Fit
Case of 10 Each for $163.62
19-071-162
View Documents
3M™
7100203203
Gray Anti-fog Blue Regular Fit
Case of 10 Each for $163.62
Description
Description
  • The adjustable ratchet temples provides angle adjustment points on the temple to help create a secure and personalized fit
  • Pressure Diffusion Temple (PDT) Technology helps diffuse pressure over the ear for a secure and comfortable fit
  • Soft touch nosepiece to help provide all-day comfort
  • Polycarbonate Lenses absorb 99.9% of UVA and UVB rays
  • Meets the requirements of ANSI Z87.1-2015 Standard as an Impact Rated Spectacle
  • 3M Catalog# SF3701, SF3702, SF3703, SF3707
Specifications
Specifications
Protective eyewear
10/Cs.
3M™ Pressure Diffusion Temple Technology
Polycarbonate
Automotive, Machining, Transportation, Sawing, Riveting
Universal
Polycarbonate
Videos
SDS
missing translation for 'documents'
missing translation for 'documents'