3M™ SecureFit™ Protective Eyewear
Pressure Diffusion Temple Technology that helps diffuse pressure over the ear to enhance frame comfort
Supplier:  3M™ 7100090435
 View more versions of this product
Catalog No. 17-623-700
Explore available promotions
Description
Description
  • Need to stock multiple frame sizes at work sites eliminated by self-adjusting feature
  • Secure, snug fit prevents slippage even during motion
  • Anti-fog coating ensures moisture free lenses in hot and humid conditions
  • Anti-scratch coating protects lenses from scratches, nicks and scrapes
  • Polycarbonate lenses absorb 99.9% UVA and UVB up to 380nm
  • 3M Catalog# SF201AF, SF201AS, SF202AF, SF202AS, SF203AF
Compliance
Meets ANSI Z87.1-2015 and CSA Z94.3 standards.
Specifications
Specifications
Anti-fog
Clear
Proprietary self-adjusting 3M Pressure Diffusion Temple Technology
ANSI Z87.1-2015, CSA Z94.3
SF201AF
Polycarbonate