3M™ SecureFit™ Protective Eyewear
Pressure Diffusion Temple Technology that helps diffuse pressure over the ear to enhance frame comfort
$3.74 - $4.33
Specifications
Certifications/Compliance ANSI Z87.1-2015, CSA Z94.3
Temple Style Proprietary self-adjusting 3M Pressure Diffusion Temple Technology
View More Specs
Products 4
Catalog Number Mfr. No. Lens Tint Price Quantity  
Catalog Number Mfr. No. Lens Tint Price Quantity  
17-623-701
View Documents
3M™
7100022210
Gray
Each for $4.04
Case of 20 Each for $80.80
17-623-702
View Documents
3M™
7100021793
Amber
Each for $4.14
Case of 20 Each for $82.80
17-623-704
View Documents
3M™
7100022211
Gray
Each for $3.74
Case of 20 Each for $74.80
17-623-700
View Documents
3M™
7100090435
Clear
Each for $4.33
Case of 20 Each for $86.60
Description
Description
  • Need to stock multiple frame sizes at work sites eliminated by self-adjusting feature
  • Secure, snug fit prevents slippage even during motion
  • Anti-fog coating ensures moisture free lenses in hot and humid conditions
  • Anti-scratch coating protects lenses from scratches, nicks and scrapes
  • Polycarbonate lenses absorb 99.9% UVA and UVB up to 380nm
  • 3M Catalog# SF201AF, SF201AS, SF202AF, SF202AS, SF203AF
Compliance
Meets ANSI Z87.1-2015 and CSA Z94.3 standards.
Specifications
Specifications
ANSI Z87.1-2015, CSA Z94.3
Proprietary self-adjusting 3M Pressure Diffusion Temple Technology
Videos
SDS
missing translation for 'documents'
missing translation for 'documents'