missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Selectophore™ Lithium Ionophore VI, Function Tested, 99.0% (HPLC), MilliporeSigma™ Supelco™
Supplier: Merck Emd Millipore 6256710MG
Specifications
106868-21-7 | |
C24H32O4 | |
MFCD00037498 | |
6,6-Dibenzyl-1,4,8-11-tetraoxacyclotetradecane; 6,6-Dibenzyl-14-crown-4 | |
C(C1=CC=CC=C1)C1(CC2=CC=CC=C2)COCCOCCCOCCOC1 | |
384.52 | |
≥99.0% (HPLC) | |
Selectophore |
Bottomless Glass Bottle with Fused Cone | |
C24H32O4 | |
10 mg | |
KDMUFHBMXZLDLI-UHFFFAOYSA-N | |
6,6-dibenzyl-1,4,8,11-tetraoxacyclotetradecane | |
384.51 | |
Selectophore™ |
Chemical Identifiers
106868-21-7 | |
384.52 | |
KDMUFHBMXZLDLI-UHFFFAOYSA-N | |
6,6-dibenzyl-1,4,8,11-tetraoxacyclotetradecane |
C24H32O4 | |
MFCD00037498 | |
6,6-Dibenzyl-1,4,8-11-tetraoxacyclotetradecane; 6,6-Dibenzyl-14-crown-4 | |
C(C1=CC=CC=C1)C1(CC2=CC=CC=C2)COCCOCCCOCCOC1 |
Selectophore is a trademark of Sigma-Aldrich Chemie GmbH