Learn More
Selenium(I) chloride, 99%, Thermo Scientific Chemicals
$785.26 - $1,285.63
Chemical Identifiers
CAS | 10025-68-0 |
---|---|
Molecular Formula | Cl2Se2 |
Molecular Weight (g/mol) | 228.84 |
MDL Number | MFCD00011226 |
InChI Key | VIEXQFHKRAHTQS-UHFFFAOYSA-N |
Synonym | diselenium dichloride, selenium monochloride, selenium chloride se2cl2, unii-2a5292fc4p, selenium i chloride, dichlorodiselenane, dichloro perselenide, se2cl2, selenium i chloride, ampuled under argon trace metals basis 25g |
PubChem CID | 66206 |
IUPAC Name | chloroselanyl selenohypochlorite |
SMILES | Cl[Se][Se]Cl |
Description
Selenium monochloride is used in electronics, instrumentation and other industrial purposes. It is used as a reagent for the synthesis of Se-containing compounds.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
10025-68-0 | |
228.84 | |
VIEXQFHKRAHTQS-UHFFFAOYSA-N | |
66206 | |
Cl[Se][Se]Cl |
Cl2Se2 | |
MFCD00011226 | |
diselenium dichloride, selenium monochloride, selenium chloride se2cl2, unii-2a5292fc4p, selenium i chloride, dichlorodiselenane, dichloro perselenide, se2cl2, selenium i chloride, ampuled under argon trace metals basis 25g | |
chloroselanyl selenohypochlorite |
Specifications
127°C (decomposition) | |
10025-68-0 | |
25 g | |
MFCD00011226 | |
14,8432 | |
Decomposes in water; Soluble in chloroform, benzene, carbon tetrachloride, carbon disulfide | |
Cl[Se][Se]Cl | |
228.84 | |
228.83 | |
Acidic | |
2.77 g/mL |
-85°C | |
Liquid | |
Cl2Se2 | |
UN2922 | |
diselenium dichloride, selenium monochloride, selenium chloride se2cl2, unii-2a5292fc4p, selenium i chloride, dichlorodiselenane, dichloro perselenide, se2cl2, selenium i chloride, ampuled under argon trace metals basis 25g | |
VIEXQFHKRAHTQS-UHFFFAOYSA-N | |
chloroselanyl selenohypochlorite | |
66206 | |
99% | |
Moisture sensitive | |
Selenium(I) chloride |
Safety and Handling
GHS H Statement
H301-H331-H373
Toxic if swallowed.
Toxic if inhaled.
May cause damage to organs through prolonged or repeated exposure.
P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c
H301+H331-H314-H335-H373
DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUIDS, TOXIC, N.O.S.
EINECSNumber : 233-037-8
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only