Globe Scientific Self-Standing Sterile Transport Tubes with Attached Screw Cap
Designed for the safe storage and transport of biological materials.
Supplier:  Globe Scientific 6102S
 View more versions of this product
Catalog No. 22-010-1224
Explore available promotions
Description
Description
  • Tubes are made of polypropylene
  • With molded graduations
  • Self-standing with conical bottom
  • Transparent tubes allows for unobstructed viewing of samples
  • Shatter-proof and may be exposed to freezing temperatures
  • Leak-resistant polyethylene screw caps can be removed or sealed in single turn
Specifications
Specifications
White
Sterile
10 mL
1 to 10 mL
Self Standing Transport Tube
Autoclavable (Vial Only)
92 mm
Polypropylene
Single-use
Yes
White Screwcap
Polyethylene
Screw Cap
Etched