missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Semicarbazide hydrochloride, 99+%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 132641000

 View more versions of this product

Catalog No. AC132641000


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Semicarbazide hydrochloride
170.0°C to 178.0°C
Authentic
Plastic bottle
H2NCONHNH2·HCl
100 g
01,1000; 15,284
semicarbazide hydrochloride, aminourea hydrochloride, semicarbazide hcl, hydrazinecarboxamide hydrochloride, hydrazinecarboxamide, monohydrochloride, semicarbazide chloride, amidourea hydrochloride, carbamylhydrazine hydrochloride, semicarbazide monohydrochloride, semicarbazide, monohydrochloride
XHQYBDSXTDXSHY-UHFFFAOYSA-N
aminourea;hydrochloride
11236
111.53
Crystalline Powder and/or Chunks
563-41-7
White
99+%
CH6ClN3O
MFCD00013009
03,98
15,8583
(10% in water) Clear colorless
[H+].[Cl-].NNC(N)=O
111.53
CHEBI:82532
99+%

Chemical Identifiers

563-41-7
111.53
XHQYBDSXTDXSHY-UHFFFAOYSA-N
11236
aminourea;hydrochloride
CH6ClN3O
MFCD00013009
semicarbazide hydrochloride, aminourea hydrochloride, semicarbazide hcl, hydrazinecarboxamide hydrochloride, hydrazinecarboxamide, monohydrochloride, semicarbazide chloride, amidourea hydrochloride, carbamylhydrazine hydrochloride, semicarbazide monohydrochloride, semicarbazide, monohydrochloride
CHEBI:82532
[H+].[Cl-].NNC(N)=O
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Suspected of causing cancer.
Causes skin irritation.
Causes serious eye irritation.
Toxic if swallowed.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
If skin irritation occurs: Get medical advice/attention.
If eye irritatio

GHS Signal Word: Danger

EINECSNumber : 209-247-

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only