Learn More
Semicarbazide hydrochloride, 99+%
CAS: 563-41-7 | CH6ClN3O | 111.53 g/mol
$53.58 - $152.46
Chemical Identifiers
| CAS | 563-41-7 |
|---|---|
| Molecular Formula | CH6ClN3O |
| Molecular Weight (g/mol) | 111.53 |
| MDL Number | MFCD00013009 |
| InChI Key | XHQYBDSXTDXSHY-UHFFFAOYSA-N |
| Synonym | semicarbazide hydrochloride, aminourea hydrochloride, semicarbazide hcl, hydrazinecarboxamide hydrochloride, hydrazinecarboxamide, monohydrochloride, semicarbazide chloride, amidourea hydrochloride, carbamylhydrazine hydrochloride, semicarbazide monohydrochloride, semicarbazide, monohydrochloride |
| PubChem CID | 11236 |
| ChEBI | CHEBI:82532 |
| IUPAC Name | aminourea;hydrochloride |
| SMILES | [H+].[Cl-].NNC(N)=O |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
AC132641000
|
Thermo Scientific Chemicals
132641000 |
100 g | Plastic bottle |
N/A
|
|
||||
|
AC132645000
|
Thermo Scientific Chemicals
132645000 |
500 g | Plastic bottle |
N/A
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 563-41-7 | |
| 111.53 | |
| XHQYBDSXTDXSHY-UHFFFAOYSA-N | |
| 11236 | |
| aminourea;hydrochloride |
| CH6ClN3O | |
| MFCD00013009 | |
| semicarbazide hydrochloride, aminourea hydrochloride, semicarbazide hcl, hydrazinecarboxamide hydrochloride, hydrazinecarboxamide, monohydrochloride, semicarbazide chloride, amidourea hydrochloride, carbamylhydrazine hydrochloride, semicarbazide monohydrochloride, semicarbazide, monohydrochloride | |
| CHEBI:82532 | |
| [H+].[Cl-].NNC(N)=O |
Specifications
| 563-41-7 | |
| White | |
| 99+% | |
| CH6ClN3O | |
| MFCD00013009 | |
| 03,98 | |
| 15,8583 | |
| (10% in water) Clear colorless | |
| [H+].[Cl-].NNC(N)=O | |
| 111.53 | |
| CHEBI:82532 | |
| 99+% | |
| Semicarbazide hydrochloride |
| 170.0°C to 178.0°C | |
| Authentic | |
| Plastic bottle | |
| H2NCONHNH2·HCl | |
| 100 g | |
| 01,1000; 15,284 | |
| semicarbazide hydrochloride, aminourea hydrochloride, semicarbazide hcl, hydrazinecarboxamide hydrochloride, hydrazinecarboxamide, monohydrochloride, semicarbazide chloride, amidourea hydrochloride, carbamylhydrazine hydrochloride, semicarbazide monohydrochloride, semicarbazide, monohydrochloride | |
| XHQYBDSXTDXSHY-UHFFFAOYSA-N | |
| aminourea;hydrochloride | |
| 11236 | |
| 111.53 | |
| Crystalline Powder and/or Chunks |
Safety and Handling
GHS H Statement
Suspected of causing cancer.
Causes skin irritation.
Causes serious eye irritation.
Toxic if swallowed.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
If skin irritation occurs: Get medical advice/attention.
If eye irritatio
GHS Signal Word: Danger
EINECSNumber : 209-247-
RUO – Research Use Only