Sensidyne™ Detector Tubes: Sulfur Compounds
Supplier:  Sensidyne™ 244U
 View more versions of this product
Catalog No. 19-122-900
Explore available promotions
Description
Description
All detector tubes are sold in packs of 10.
Specifications
Specifications
Sulfuric Acid
5
100 sec.
5° to 40°C
0.5 to 5 mg/m3
10
41°F to 104°F
Product Suggestions
Product Suggestions