United Scientific™ Separatory Funnel DFS Item
Separatory Funnels provide excellent clarity and strength.
$84.32 - $86.35
Specifications
Material Polypropylene
Color Clear
Product Type Pear-shaped Separatory Funnel
View More Specs
Products 2
Catalog Number Mfr. No. Capacity (Metric) Price Quantity  
Catalog Number Mfr. No. Capacity (Metric) Price Quantity  
S90334B DFS Item
View Documents
United Scientific™
81202
250 mL
Each for $84.32
S90334C DFS Item
View Documents
United Scientific™
81203
500 mL
Each for $86.35
Description
Description
  • These pear-shaped polypropylene separating funnels feature excellent clarity and strength, and include a leak-proof threaded screw cap
  • The polypropylene stopcock is fitted with a self-lubricating PTFE plug
  • Not graduated
Specifications
Specifications
Polypropylene
Clear
Pear-shaped Separatory Funnel
Videos
SDS
missing translation for 'documents'
missing translation for 'documents'