United Scientific™ Separatory Funnel
Separatory Funnels provide excellent clarity and strength.
Supplier: United Scientific™ 81202
Description
- These pear-shaped polypropylene separating funnels feature excellent clarity and strength, and include a leak-proof threaded screw cap
- The polypropylene stopcock is fitted with a self-lubricating PTFE plug
- Not graduated
Specifications
75 mm | |
Polypropylene | |
45 mm |
250 mL | |
Clear | |
Pear-shaped Separatory Funnel |