missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Honeywell™ Servus™ Economy PVC Knee Boots

Graduated boot heights

$39.04 - $39.04

Specifications

Color Black
For Use With (Application) Agriculture, construction, food services, municipal services, oil and Gas, pharmaceutical, transportation, utilities
Height (English) 16 in.
Material PVC
Seam Type Seamless
View More Specs

Products 1
Catalog Number Mfr. No. Certifications/Compliance Size Toe Style Price Quantity  
Catalog Number Mfr. No. Certifications/Compliance Size Toe Style Price Quantity  
29-636-3168
View Documents
Honeywell™
18821C10
ASTM F2413-11 M I/75 C/75, ASTM F2413-05 10 Steel Toe
Pair for $39.04
 
Description

Description

  • Two-stage injection-molded PVC construction
  • Scalloped top line edge design and graduated boot heights accommodate flexing in the calf
  • Trac10™ outsole enhances traction and durability with an ergonomic frame design
  • Removable EVA Foot Form insole provides comfort
  • Reinforced at critical stress points
  • Style 18821 features Safe-Toe™ steel toe cap
  • 16 in. high (41cm)
  • Black

Compliance

Style 18821 boots with steel toe meet ASTM™ F2413-11 M I/75 C/75

Specifications

Specifications

Black
16 in.
Seamless
Unique trac10™ outsole, contour cushion insoles
Agriculture, construction, food services, municipal services, oil and Gas, pharmaceutical, transportation, utilities
PVC
Steel
Yes
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'