Epredia™ Cytoblock™ Cell Block Preparation System
Facilitate quick, reliable processing of FNAs, tissue fragments, and cell buttons with the Cytoblock™ Cell Block Preparation System.
Supplier:  Epredia™ 7401150
 View more versions of this product
Catalog No. 74-011-50
Explore available promotions
Description
Description
  • Increases ability to process scanty or unseen material for cell block and reduces loss of critical material
  • Suitable for immunohistochemical techniques
  • Replaces time-consuming and costly agar and thrombin techniques
  • Requires phosphate-free fixative on processor, such as Epredia™ fixatives Zinc Formal-Fixx™, Formal-Fixx, or Glyo-Fixx™ (formalin substitute) or simply bypass formalin fixation steps on processor
  • Cytoblock system can also be run on a rapid biopsy program
Specifications
Specifications
Cell Preparation System
Cell Block Preparation System