Learn More
Epredia™ Cytoblock™ Cell Block Preparation System
Facilitate quick, reliable processing of FNAs, tissue fragments, and cell buttons with the Cytoblock™ Cell Block Preparation System.
$649.38 - $649.38
Products 1
Catalog Number Mfr. No. Type Price Quantity  
Catalog Number Mfr. No. Type Price Quantity  
74-011-50
View Documents
Epredia™
7401150
Cell Preparation System
Each for $649.38
Only null left
Description
Description
  • Increases ability to process scanty or unseen material for cell block and reduces loss of critical material
  • Suitable for immunohistochemical techniques
  • Replaces time-consuming and costly agar and thrombin techniques
  • Requires phosphate-free fixative on processor, such as Epredia™ fixatives Zinc Formal-Fixx™, Formal-Fixx, or Glyo-Fixx™ (formalin substitute) or simply bypass formalin fixation steps on processor
  • Cytoblock system can also be run on a rapid biopsy program
Videos
SDS
missing translation for 'documents'
missing translation for 'documents'