Epredia™ Cytospin™ Collection Fluid
Store specimens or unstained slides for weeks without adverse effect with Epredia™ Shandon™ Cytospin™ Collection Fluid.
$138.06 - $1100.41
Specifications
Product Type Collection Fluid
Solution Type Alcohol/Carbowax (PEG) combination
View More Specs
Products 4
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
9990323
View Documents
Epredia™
9990323
120mL
Case of 125 Each for $941.03
6768315
View Documents
Epredia™
6768315
500mL
Case of 2 Each for $138.06
6768001
View Documents
Epredia™
6768001
4 L
Each for $226.65
28-601-79
View Documents
Epredia™
9990310
10L
Case of 2 Each for $1,100.41
Description
Description
  • Packaged in convenient pump bottle or pre-filled specimen cups
  • Ready-to-use alcohol/Carbowax™ (PEG) combination for direct specimen collection and fixation
Specifications
Specifications
Collection Fluid
Alcohol/Carbowax (PEG) combination
Videos
SDS
missing translation for 'documents'
missing translation for 'documents'