Epredia™ Shandon™ Eosin-Y Stain
Produce brilliant staining with Epredia™ Shandon™ Eosin-Y, a fast-acting cytoplasmic counterstain.
Supplier:  Epredia™ 6766008
 View more versions of this product
Catalog No. 28-600-90
Explore available promotions
Description
Description
  • Ready-to-use
  • Produces consistent results
  • Stable and precipitate-free; no filtering
  • Compatible with all alum hematoxylin stains
  • Available in alcoholic or aqueous formulations
Specifications
Specifications
Alcoholic Eosin Y Stain
4L