Epredia™ Shandon™ Eosin-Y Stain
Produce brilliant staining with Epredia™ Shandon™ Eosin-Y, a fast-acting cytoplasmic counterstain.
$333.35 - $444.41
Products 2
Catalog Number Mfr. No. Type Quantity Price Quantity  
Catalog Number Mfr. No. Type Quantity Price Quantity  
28-600-86
View Documents
Epredia™
6766007
Alcoholic Eosin Y Stain 1L
Case of 2 Each for $333.35
28-600-90
View Documents
Epredia™
6766008
Alcoholic Eosin Y Stain 4L
Case for $444.41
Description
Description
  • Ready-to-use
  • Produces consistent results
  • Stable and precipitate-free; no filtering
  • Compatible with all alum hematoxylin stains
  • Available in alcoholic or aqueous formulations
SDS
missing translation for 'documents'
missing translation for 'documents'