missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Epredia™ Shandon™ Round Autopsy Saw Blade

Epredia™ Shandon Round Autopsy Saw Blades are available with or without arbor.

Supplier:  Epredia™ 1002103

 View more versions of this product

Catalog No. 38-103-69

Explore available promotions

Description

Description

  • Blade diameter: 2.5 in. (6.3cm) Cutting depth: 1.75 in. (44mm)
  • Permanently bonded stainless-steel arbor (10004 only)
Specifications

Specifications

No
1.75 in.
2.5 in.
Round Autopsy Saw
44 mm
6.3 cm
Videos
SDS
Documents

Documents

Promotions

Promotions