Epredia™ Signature Series™ Clear-Rite 3™
Render complete deparaffinization and clearing during the staining process with Epredia™ Signature Series Clear-Rite 3.
Supplier:  Epredia™ 6901
 View more versions of this product
Catalog No. 22-046-341
Explore available promotions
Includes: (4) 1-gallon bottles
Description
Description
  • Very low odor; Benzene-free
  • Use in all tissue processors and automatic stainers
  • Does not produce hard or brittle specimens, even after prolonged exposure
Specifications
Specifications
Clearing Agent
4 x 1 gal.