Epredia™ Signature Series™ Clear-Rite 3™
Render complete deparaffinization and clearing during the staining process with Epredia™ Signature Series Clear-Rite 3.
$515.00 - $897.94
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
22-046-341
|
Epredia™
6901 |
4 x 1 gal. |
Case of 4 Each for $515.00
|
|
|||||
69-15
|
Epredia™
SH6915 |
5 Liter |
Case of 4 Each for $897.94
|
|
|||||
Description
- Very low odor; Benzene-free
- Use in all tissue processors and automatic stainers
- Does not produce hard or brittle specimens, even after prolonged exposure
Specifications
Clearing Agent |