Epredia™ Signature Series™ Clear-Rite 3™
Render complete deparaffinization and clearing during the staining process with Epredia™ Signature Series Clear-Rite 3.
Supplier: Epredia™ SH6915
Description
- Very low odor; Benzene-free
- Use in all tissue processors and automatic stainers
- Does not produce hard or brittle specimens, even after prolonged exposure
Specifications
5 Liter |