Epredia™ Signature Series™ Flex™ Dehydrating Reagent
Achieve brilliant cytoplasmic staining and complete dehydration with Epredia™ Signature Series Flex™ 100, 95, 80.
Supplier: Epredia™ 8101
Description
- Patented formulation of methyl and isopropyl alcohol minimizes over-dehydration in smaller biopsies
- Excellent for lipid extraction allowing for increased paraffin infiltration
- May be used as a solvent for special stains
Specifications
Flex 100 Dehydrating Reagent | |
1 gal. |