Epredia™ Signature Series™ Flex™ Dehydrating Reagent
Achieve brilliant cytoplasmic staining and complete dehydration with Epredia™ Signature Series Flex™ 100, 95, 80.
$182.88 - $981.68
Catalog Number | Mfr. No. | Quantity | Product Type | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Product Type | Price | Quantity | ||||
22-046344
|
Epredia™
8101 |
1 gal. | Flex 100 Dehydrating Reagent |
Case of 4 Each for $182.88
|
|
||||
22-046346
|
Epredia™
8201 |
1 gal. | Flex 95 Dehydrating Reagent |
Case of 4 Each for $223.29
|
|
||||
82-15
|
Epredia™
8215 |
1.32 gal. | Flex 95 Dehydrating Reagent |
Case of 4 Each for $645.05
|
|
||||
22-046348
|
Epredia™
8301R |
1 gal. | Flex 80 Dehydrating Reagent |
Case of 4 Each for $415.82
|
|
||||
83-15
|
Epredia™
8315 |
1.32 gal. | Flex 80 Dehydrating Reagent |
Case of 4 Each for $981.68
|
|
||||
Description
- Patented formulation of methyl and isopropyl alcohol minimizes over-dehydration in smaller biopsies
- Excellent for lipid extraction allowing for increased paraffin infiltration
- May be used as a solvent for special stains