Epredia™ Signature Series™ Flex™ Dehydrating Reagent
Achieve brilliant cytoplasmic staining and complete dehydration with Epredia™ Signature Series Flex™ 100, 95, 80.
Supplier:  Epredia™ 8215
 View more versions of this product
Catalog No. 82-15
Explore available promotions
Description
Description
  • Patented formulation of methyl and isopropyl alcohol minimizes over-dehydration in smaller biopsies
  • Excellent for lipid extraction allowing for increased paraffin infiltration
  • May be used as a solvent for special stains
Specifications
Specifications
Flex 95 Dehydrating Reagent
1.32 gal.