SiliCycle™ SiliaPrep™ SPE Cartridges - Silica
SPE Cartridges packed with ungrafted silica, for extraction of various compounds from non-polar solvents.
Supplier:  SiliCycle™ SPER10030B06J
 View more versions of this product
Catalog No. 01184648
Explore available promotions
Description
Description
  • For use with vacuum manifolds and automated SPE equipments
  • Tightly packed and homogeneous silica bed for reliability and increased productivity
  • Tight particle size distribution and very good packing with no fines, allowing excellent separation
  • High recoveries and yields
  • Reproducible flow rates lot-to-lot
  • Flanged polypropylene tubes
  • 20 μm polyethylene frits
Specifications
Specifications
Silica
Extraction of various polar compounds from non-polar solvents
40 to 63 μm
Bare Silica (Silicon Dioxyde, SiO2)
6 mL
Irregular
50 Pk.
12.7 x 77.8 mm
12.7 mm
300 mg
Vacuum Manifolds or Automated SPE Equipment
SiliaPrep
SPE Cartridge
Polypropylene (PP) tube, Polyethylene (PE) frits
60 Å
500 m2/g
77.8 mm
SDS
Documents
Documents
Promotions
Promotions
Keep dry