missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing
Silica Gel Desiccant (6-14 Mesh, Certified ACS), Fisher Chemical™
Chemical Identifiers
CAS |
63231-67-4 |
Molecular Formula |
H2O3Si |
Molecular Weight (g/mol) |
78.098 |
MDL Number |
MFCD00011232 |
InChI Key |
IJKVHSBPTUYDLN-UHFFFAOYSA-N |
PubChem CID |
14768 |
IUPAC Name |
dihydroxy(oxo)silane |
SMILES |
O[Si](=O)O |
View More Specs
Products
1
Catalog Number |
Mfr. No. |
Quantity |
Packaging |
Price |
Quantity |
|
|
|
|
S684-212
SDS
View Documents
|
Fisher Chemical
S684212
|
2.5 kg |
Poly Bottle |
|
|
Add to cart
|
|
|
---|
|
Description
Specifications
Chemical Identifiers
|
63231-67-4 |
|
78.098 |
|
IJKVHSBPTUYDLN-UHFFFAOYSA-N |
|
dihydroxy(oxo)silane |
|
H2O3Si |
|
MFCD00011232 |
|
14768 |
|
O[Si](=O)O |
Specifications
|
1710°C |
|
Solid |
|
2.3 to 7.4 |
|
2.1g/cm³ |
|
2.5 kg |
|
H2O3Si |
|
6 to 14 Mesh |
|
IJKVHSBPTUYDLN-UHFFFAOYSA-N |
|
dihydroxy(oxo)silane |
|
14768 |
|
Certified ACS |
|
2230°C |
|
White |
|
63231-67-4 |
|
Silica Gel |
|
Pass Test |
|
MFCD00011232 |
|
Poly Bottle |
|
O[Si](=O)O |
|
78.098 |
|
Pass Test |
|
Silica Gel Desiccant, 6-14 Mesh |
Videos
Safety and Handling
SDS
missing translation for 'documents'