missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Silica Gel Desiccant (6-14 Mesh, Certified ACS), Fisher Chemical™

Supplier:  Fisher Chemical S684212

 View more versions of this product

Catalog No. S684-212



Explore available promotions

Description

Description

Specifications

Specifications

1710°C
Solid
2.3 to 7.4
6-14 Mesh
2.1g/cm³
2.5 kg
H2O3Si
6 to 14 Mesh
IJKVHSBPTUYDLN-UHFFFAOYSA-N
dihydroxy(oxo)silane
14768
Certified ACS
2230°C
White
Silica Gel Desiccant
63231-67-4
Silica Gel
Pass Test
MFCD00011232
Poly Bottle
O[Si](=O)O
78.098
Pass Test

Chemical Identifiers

63231-67-4
78.098
IJKVHSBPTUYDLN-UHFFFAOYSA-N
dihydroxy(oxo)silane
H2O3Si
MFCD00011232
14768
O[Si](=O)O
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

SDS Documents

Documents

Promotions

Promotions