missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Silicagel orange, for drying purposes, non toxic grade, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 392030050
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Drying agentSpecifications
Beads | |
Silica gel orange | |
Silica Gel | |
Al2O5Si | |
MFCD00077645 | |
Solubility in water: insoluble. | |
162.04 |
Orange | |
1327-36-2 | |
5 kg | |
2% max. (120°C, 2 h) | |
Plastic Drum | |
HNPSIPDUKPIQMN-UHFFFAOYSA-N |
Chemical Identifiers
1327-36-2 | |
162.04 | |
HNPSIPDUKPIQMN-UHFFFAOYSA-N |
Al2O5Si | |
MFCD00077645 |
RUO – Research Use Only