missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Silicagel orange, for drying purposes, non toxic grade, Thermo Scientific Chemicals
$80.80 - $715.67
Chemical Identifiers
CAS | 1327-36-2 |
---|---|
Molecular Formula | Al2O5Si |
Molecular Weight (g/mol) | 162.04 |
MDL Number | MFCD00077645 |
InChI Key | HNPSIPDUKPIQMN-UHFFFAOYSA-N |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AC392032500
|
Thermo Scientific Chemicals
392032500 |
250 g | Glass Bottle |
Each for $80.80
|
|
||||
AC392030010
|
Thermo Scientific Chemicals
392030010 |
1 kg | Glass Bottle |
Each for $178.74
|
|
||||
AC392030050
|
Thermo Scientific Chemicals
392030050 |
5 kg | Plastic Drum |
Each for $715.67
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Drying agentChemical Identifiers
1327-36-2 | |
162.04 | |
HNPSIPDUKPIQMN-UHFFFAOYSA-N |
Al2O5Si | |
MFCD00077645 |
Specifications
Beads | |
1327-36-2 | |
250 g | |
2% max. (120°C, 2 h) | |
Glass Bottle | |
HNPSIPDUKPIQMN-UHFFFAOYSA-N | |
Silica gel orange |
Orange | |
Silica Gel | |
Al2O5Si | |
MFCD00077645 | |
Solubility in water: insoluble. | |
162.04 |
RUO – Research Use Only