Learn More
Silicon dioxide, 99.998%, (trace metal basis), Thermo Scientific Chemicals
For ultra trace analysis
$77.38 - $714.48
Chemical Identifiers
CAS | 14808-60-7 |
---|---|
Molecular Formula | O2Si |
Molecular Weight (g/mol) | 60.08 |
MDL Number | MFCD00011232,MFCD00217788,MFCD00163736,MFCD07370733 |
InChI Key | VYPSYNLAJGMNEJ-UHFFFAOYSA-N |
Synonym | silica, silicon dioxide, quartz, cristobalite, diatomaceous earth, tridymite, silicic anhydride, aerosil, infusorial earth, silica, amorphous |
PubChem CID | 24261 |
ChEBI | CHEBI:30563 |
IUPAC Name | silanedione |
SMILES | O=[Si]=O |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AC437150050
|
Thermo Scientific Chemicals
437150050 |
5 g | Glass bottle |
Each for $77.38
|
|
||||
AC437150200
|
Thermo Scientific Chemicals
437150200 |
20 g | Glass bottle |
Each for $224.74
|
|
||||
AC437151000
|
Thermo Scientific Chemicals
437151000 |
100 g | Glass bottle |
Each for $714.48
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For ultra trace analysisChemical Identifiers
14808-60-7 | |
60.08 | |
VYPSYNLAJGMNEJ-UHFFFAOYSA-N | |
24261 | |
silanedione |
O2Si | |
MFCD00011232,MFCD00217788,MFCD00163736,MFCD07370733 | |
silica, silicon dioxide, quartz, cristobalite, diatomaceous earth, tridymite, silicic anhydride, aerosil, infusorial earth, silica, amorphous | |
CHEBI:30563 | |
O=[Si]=O |
Specifications
1610.0°C | |
Colorless to Yellow | |
5 g | |
O2Si | |
MFCD00011232,MFCD00217788,MFCD00163736,MFCD07370733 | |
Solubility in water: insoluble | |
O=[Si]=O | |
60.08 | |
CHEBI:30563 | |
99.998% | |
Glass bottle |
14808-60-7 | |
Solution | |
44.5 to 47.9% (Si) | |
SiO2 | |
silica, silicon dioxide, quartz, cristobalite, diatomaceous earth, tridymite, silicic anhydride, aerosil, infusorial earth, silica, amorphous | |
VYPSYNLAJGMNEJ-UHFFFAOYSA-N | |
silanedione | |
24261 | |
60.08 | |
Trace Metal Basis | |
Silicon dioxide |
Safety and Handling
GHS H Statement
May cause cancer by inhalation.
May cause damage to organs through prolonged or repeated exposure.
GHS P Statement
Obtain special instructions before use.
Wear protective gloves/protective clothing/eye protection/face protection.
IF exposed or concerned: Get medical advice/attention.
GHS Signal Word: Warning
EINECSNumber : 238-878-4
RUO – Research Use Only