Learn More
Silver acetate, 99%, pure, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 202391000
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Suitable for battery materials development.
Specifications
Silver acetate | |
563-63-3 | |
Powder | |
99% | |
CH3CO2Ag | |
01,1002; 03,362; 06,511; 08,440; 12,256; 13,285 | |
silver acetate, silver i acetate, silver 1+ acetate, silver monoacetate, unii-19pps85f9h, acetic acid, silver 1+ salt, acetic acid silver salt, silver 1+ ion acetate, acetic acid, silver 1+ salt 1:1, silver acetate ag oac | |
CQLFBEKRDQMJLZ-UHFFFAOYSA-M | |
silver;acetate | |
11246 | |
99% | |
Authentic | |
3.25 |
99% | |
Beige-Gray to White | |
100 g | |
C2H3AgO2 | |
MFCD00012446 | |
15,8644 | |
Solubility in water: 10.2g/L (20°C). Other solubilities: soluble in nitric acid | |
CC(=O)[O-].[Ag+] | |
166.91 | |
166.91 | |
Pure | |
Glass bottle | |
3.2500g/mL |
Chemical Identifiers
563-63-3 | |
166.91 | |
CQLFBEKRDQMJLZ-UHFFFAOYSA-M | |
11246 | |
CC(=O)[O-].[Ag+] |
C2H3AgO2 | |
MFCD00012446 | |
silver acetate, silver i acetate, silver 1+ acetate, silver monoacetate, unii-19pps85f9h, acetic acid, silver 1+ salt, acetic acid silver salt, silver 1+ ion acetate, acetic acid, silver 1+ salt 1:1, silver acetate ag oac | |
silver;acetate |
Safety and Handling
GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Very toxic to aquatic life with long lasting effects.
Causes skin irritation.
GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
Avoid release to the environment.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Warning
EINECSNumber : 209-254-9
RTECSNumber : AJ4100000
TSCA : TSCA
RUO – Research Use Only