missing translation for 'onlineSavingsMsg'
Learn More

Silver acetate, 99%, pure

CAS: 563-63-3 | C2H3AgO2 | 166.91 g/mol

$363.62 - $4200.88

Chemical Identifiers

CAS 563-63-3
Molecular Formula C2H3AgO2
Molecular Weight (g/mol) 166.91
MDL Number MFCD00012446
InChI Key CQLFBEKRDQMJLZ-UHFFFAOYSA-M
Synonym silver acetate, silver i acetate, silver 1+ acetate, silver monoacetate, unii-19pps85f9h, acetic acid, silver 1+ salt, acetic acid silver salt, silver 1+ ion acetate, acetic acid, silver 1+ salt 1:1, silver acetate ag oac
PubChem CID 11246
IUPAC Name silver;acetate
SMILES CC(=O)[O-].[Ag+]
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AC202390250
SDS View Documents
Thermo Scientific Chemicals
202390250
25 g
Each for $363.62
Only null left
 
AC202391000
SDS missing translation for 'viewDocuments'
Thermo Scientific Chemicals
202391000
100 g
Each for $932.34
Only null left
 
AC202395000
SDS missing translation for 'viewDocuments'
Thermo Scientific Chemicals
202395000
500 g
Each for $4,200.88
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

Specifications

Chemical Identifiers

563-63-3
166.91
CQLFBEKRDQMJLZ-UHFFFAOYSA-M
11246
CC(=O)[O-].[Ag+]
C2H3AgO2
MFCD00012446
silver acetate, silver i acetate, silver 1+ acetate, silver monoacetate, unii-19pps85f9h, acetic acid, silver 1+ salt, acetic acid silver salt, silver 1+ ion acetate, acetic acid, silver 1+ salt 1:1, silver acetate ag oac
silver;acetate

Specifications

563-63-3
Powder
99%
CH3CO2Ag
01,1002; 03,362; 06,511; 08,440; 12,256; 13,285
silver acetate, silver i acetate, silver 1+ acetate, silver monoacetate, unii-19pps85f9h, acetic acid, silver 1+ salt, acetic acid silver salt, silver 1+ ion acetate, acetic acid, silver 1+ salt 1:1, silver acetate ag oac
CQLFBEKRDQMJLZ-UHFFFAOYSA-M
silver;acetate
11246
99%
Authentic
3.25
Silver acetate, 99%
Beige-Gray to White
25 g
C2H3AgO2
MFCD00012446
15,8644
Solubility in water: 10.2g/L (20°C). Other solubilities: soluble in nitric acid
CC(=O)[O-].[Ag+]
166.91
166.91
Pure
Glass bottle
3.2500g/mL
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Very toxic to aquatic life with long lasting effects.
Causes skin irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
Avoid release to the environment.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 209-254-9

RTECSNumber : AJ4100000

TSCA : TSCA

SDS
Documents

Documents

RUO – Research Use Only