missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Silver acetate, 99%, pure, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 202395000

 View more versions of this product

Catalog No. AC202395000


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

Specifications

Specifications

Silver acetate
563-63-3
Powder
99%
CH3CO2Ag
01,1002; 03,362; 06,511; 08,440; 12,256; 13,285
silver acetate, silver i acetate, silver 1+ acetate, silver monoacetate, unii-19pps85f9h, acetic acid, silver 1+ salt, acetic acid silver salt, silver 1+ ion acetate, acetic acid, silver 1+ salt 1:1, silver acetate ag oac
CQLFBEKRDQMJLZ-UHFFFAOYSA-M
silver;acetate
11246
99%
Authentic
3.2500g/mL
99%
Beige-Gray to White
500 g
C2H3AgO2
MFCD00012446
15,8644
Solubility in water: 10.2g/L (20°C). Other solubilities: soluble in nitric acid
CC(=O)[O-].[Ag+]
166.91
166.91
Pure
3.25

Chemical Identifiers

563-63-3
166.91
CQLFBEKRDQMJLZ-UHFFFAOYSA-M
11246
CC(=O)[O-].[Ag+]
C2H3AgO2
MFCD00012446
silver acetate, silver i acetate, silver 1+ acetate, silver monoacetate, unii-19pps85f9h, acetic acid, silver 1+ salt, acetic acid silver salt, silver 1+ ion acetate, acetic acid, silver 1+ salt 1:1, silver acetate ag oac
silver;acetate
Safety and Handling

Safety and Handling

GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Very toxic to aquatic life with long lasting effects.
Causes skin irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
Avoid release to the environment.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 209-254-9

RTECSNumber : AJ4100000

TSCA : TSCA

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only