missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Silver chromate, 99% min, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 04010722

 View more versions of this product

Catalog No. AA4010722


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Silver chromate
Powder
Ag2CrO4
UN1479
silver chromate, unii-zj5092lwu9, silver chromate vi, silberchromat, silver i chromate, 2ag.cro4, chromic acid h2cro4 , disilver 1+ salt, chromic acid h2cro4 , silver 1+ salt 1:2, chromic acid, disilver i salt, disilver dioxido dioxo chromium
OJKANDGLELGDHV-UHFFFAOYSA-N
disilver;dioxido(dioxo)chromium
62666
≤99%
Light sensitive
7784-01-2
100 g
MFCD00003402
14,8510
Slightly soluble in water; Soluble in HNO3 and ammonia
[O-][Cr](=O)(=O)[O-].[Ag+].[Ag+]
331.73
331.73
Odorless
5.63 g/mL

Chemical Identifiers

7784-01-2
331.73
OJKANDGLELGDHV-UHFFFAOYSA-N
62666
[O-][Cr](=O)(=O)[O-].[Ag+].[Ag+]
Ag2CrO4
MFCD00003402
silver chromate, unii-zj5092lwu9, silver chromate vi, silberchromat, silver i chromate, 2ag.cro4, chromic acid h2cro4 , disilver 1+ salt, chromic acid h2cro4 , silver 1+ salt 1:2, chromic acid, disilver i salt, disilver dioxido dioxo chromium
disilver;dioxido(dioxo)chromium
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H350-H272-H317
May cause cancer.
May intensify fire
oxidizer.
May cause an allergic skin reaction.

P201-P202-P210-P220-P221-P261-P272-P280g-P281-P302+P352-P308+P313-P333+P313-P363-P370+P378q-P501c

H272-H317-H350i

DOTInformation : Transport Hazard Class: 5.1; Packing Group: III; Proper Shipping Name: OXIDIZING SOLID, N.O.S.

EINECSNumber : 232-043-8

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only